Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Croscarmellose Sodium, NF, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
74811-65-7
74811-65-7

C1366, 74811-65-7

Spectrum™ Croscarmellose Sodium, NF is used commonly as a FDA-approved disintegrant in pharmaceutical manufacturing. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 14.0-28%
Loss on Drying 10%
Quantity 2.5 kg
Packaging Poly Pail
CAS Max % 100%