Learn More
Moore Medical CrossTex™ Surgical Tie-On Facemasks
Fluid resistant outer and inner layers
Manufacturer:  Moore Medical 11017
Catalog No. 19-815-348
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Description
Description
  • Extra large panel for surgical procedures
  • Extra long laces for a secure fit
  • BFE >98% at 1 micron
  • Color: Blue
  • Sold 50/box
Compliance
Meets En14683 Rating - Type IIR Standard
Specifications
Specifications
EN 14683
99.2% (PFE), 98% (BFE)
Tie-on loops
Glass-free, lint-free with fluid resistant outer layer with white fluid resistant inner layer
Blue
Yes
Extra large panel with extra long laces