Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotamiton, BP, 96-102%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

C2641, 483-63-6, C13H17NO

Spectrum™ Crotamiton, BP is used to treat scabies skin infections. All Spectrum Chemical BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Sulfated Ash 0.001
Refractive Index 1.5-1.542
Quantity 25 kg
CAS 483-63-6
CAS Max % 1
Molecular Formula C13H17NO
InChI Key DNTGGZPQPQTDQF-XBXARRHUSA-N
SMILES CCN(C(=O)\C=C\C)C1=CC=CC=C1C
IUPAC Name (2E)-N-ethyl-N-(2-methylphenyl)but-2-enamide
Molecular Weight (g/mol) 203.29
Percent Purity 96 to 102%
Show More Show Less