Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Croton Oil, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
Steel Pail
8001-28-3
8001-28-3

CR132, 8001-28-3

Spectrum™ Croton Oil is an oil prepared from the seeds of the tree, Croton tiglium. Croton oil is the source of the organic compound phorbol and is the basis of rejuvenating chemical peels, due to the caustic exfoliating effects it has on the dermal components of the skin. Used in conjunction with phenol solutions, it results in an intense reaction which leads to initial skin sloughing and then eventual regeneration. In traditional Chinese medicine it is used as an ingredient in some liniments.
Refractive Index 1.4760-1.4780
Quantity 500 g
Packaging Amber Glass Bottle
CAS Max % 100%
UN Number UN2810