Learn More
Crotonaldehyde Antibody (2D3.1), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP261922IR
Catalog No. NBP261922IR
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Crotonaldehyde Monoclonal specifically detects Crotonaldehyde in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Crotonaldehyde
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH)
0.1 mL
Specific for Crotonaldehyde modified proteins. Does not cross-react with Acrolein, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Store at 4C in the dark.
IgG1
Western Blot, ELISA
2D3.1
Western Blot, ELISA
Mouse
Protein G purified
Primary
Mouse
Purified
Recommendations
Recommendations