Learn More
Crotonaldehyde Monoclonal Antibody (2D3.1), PerCP, Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA545855
Encompass
Catalog No. PIMA545855
Add to Cart
Description
Description
Crotonaldehyde Monoclonal antibody specifically detects Crotonaldehyde in Chemical samples. It is validated for Dot Blot, ELISA, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Crotonaldehyde
Monoclonal
1 mg/mL
2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4
Mouse
Protein G
RUO
4° C
IgG1
Dot Blot, ELISA, Western Blot
2D3.1
PerCP
CRA; CRA-lysine adduct; Croton aldehyde; Crotonaldehyde
Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH)
100 μg
Primary
Liquid
Chemical
Recommendations
Recommendations