Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotonaldehyde Mouse anti-Chemical, Clone: 2A8.1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527578

Catalog No. PIMA527578


Add to cart

Description

Description

Crotonaldehyde is a chemical compound that is an unsaturated aldehyde, used as a preservative and found in food such as soybean oil.
Specifications

Specifications

Crotonaldehyde
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Crotonaldehyde, Croton aldehyde, Crotonaldehyde (CRA), CRA, Crotonaldehyde conjugated protein, Crotonaldehyde modified protein, Crotonaldehyde-conjugated protein, Crotonaldehyde-modified protein, CRA-lysine adduct, Crotonaldehyde-lysine adduct
Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH)
Protein G
RUO
Antibody
Monoclonal
ELISA, Western Blot
2A8.1
Unconjugated
Liquid
Mouse
IgG1
100 μg
-20°C
Primary
Chemical
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit