Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crotonaldehyde Monoclonal Antibody (2D3.1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527552
Description
Crotonaldehyde Monoclonal antibody specifically detects Crotonaldehyde in Chemical samples. It is validated for ELISA, Western Blot.Specifications
Crotonaldehyde | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
-20°C | |
Liquid | |
Chemical |
ELISA, Western Blot | |
2D3.1 | |
Unconjugated | |
CRA; CRA-lysine adduct; Croton aldehyde; Crotonaldehyde | |
Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH) | |
100 μg | |
Primary | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction