Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crotonaldehyde Mouse anti-Mouse, FITC, Clone: 2D3.1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261922F
Description
Crotonaldehyde Monoclonal antibody specifically detects Crotonaldehyde in Mouse samples. It is validated for ELISA.Specifications
Crotonaldehyde | |
2D3.1 | |
ELISA | |
Purified | |
Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH) | |
Protein G purified | |
Store at 4C in the dark. | |
Monoclonal | |
Mouse |
ELISA | |
FITC | |
PBS with 0.05% Sodium Azide | |
Mouse | |
IgG1 | |
0.1 ml | |
Primary | |
Specific for Crotonaldehyde modified proteins. Does not cross-react with Acrolein, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title