Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotyl chloride, 70%, tech., ACROS Organics™

$181.40 - $672.90

Specifications

Chemical Name or Material Crotyl chloride
Name Note tech., 70%
CAS 591-97-9, 563-52-0, 1190-22-3
Grade Technical
Assay Percent Range 65% min. (GC)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC154805000
View Documents
Acros Organics
154805000
500mL Glass bottle
Each for $672.90
Order in Bulk

Request bulk or custom formats

Add to cart
 
AC154801000
View Documents
Acros Organics
154801000
100mL Glass bottle
Each for $181.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Crotyl chloride
591-97-9, 563-52-0, 1190-22-3
65% min. (GC)
CH3CH=CHCH2Cl
MFCD00000987
tech., 70%
Technical
C4H7Cl
90.55
1-Chloro-2-butene
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-739-5

RTECSNumber : EM4264000

TSCA : TSCA

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit