Learn More
Crry/p65 Mouse, anti-Mouse, R718, Clone: 1F2, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752234
Catalog No. BDB752234
Add to cart
Description
Description
The 1F2 mAb reacts with Crry/p65, a cell-surface protein that blocks complement activation and C3 deposition by both the classical and alternative pathways. Based on nucleotide sequence identity, Crry/p65 is the genetic homologue of human CR1 (Complement Receptor 1). However, it has been proposed that Crry/p65 is the functional homologue of human MCP (Membrane Cofactor Protein) and/or DAF (Decay-Accelerating Factor), due to the similar functions these proteins share and the fact that Crry/p65 does not display CR1's complement-receptor activity. Crry/p65 is expressed in most organs, such as thymus, kidney, uterus, skin, lung, pancreas, heart, stomach, large and small intestine, and spleen. Platelets, brain, and muscle express low, but detectable levels of Crry/p65. In addition, Crry/p65 is expressed in J774 cells, bone marrow-derived macrophages, and thioglycollate-elicited peritoneal macrophages, as determined by RT-PCR. Crry is crucial in the regulation of complement activation during early mouse embryonic development and essential in fetomaternal tolerance. The 1F2 mAb partially blocks Crry/p65-mediated complement protection in vitro.
Specifications
Specifications
Crry/p65
1F2
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Mouse