Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crry/p65 Mouse anti-Rat, BV421, Clone: 512, BD OptiBuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 744822
Description
Crry/p65 Monoclonal antibody specifically detects Crry/p65 in Rat samples. It is validated for Flow Cytometry.Specifications
Crry/p65 | |
512 | |
Brilliant Violet 421 | |
Mouse | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG1, κ | |
RUO | |
Primary | |
Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title