Learn More
Abnova™ CRSP7 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00009441T02
Catalog No. 89-940-588
Add to cart
Specifications
Specifications
Western Blot
293T
NM_004831.3
pCMV-CRSP7 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
CRSP7 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 65.4kDa
Transient overexpression cell lysate was tested with anti-DFFA antibody (Mfr. No. H00009441-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)