Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTAM Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0204-21A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212208AF532

Catalog No. NP212208A32


Add to cart

Description

Description

CRTAM Monoclonal antibody specifically detects CRTAM in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications

Specifications

CRTAM
MM0204-21A9
Western Blot, Flow Cytometry
CD355 antigen, class I MHC restricted T cell associated molecule, class-I MHC-restricted T cell associated molecule, Class-I MHC-restricted T-cell-associated molecule, cytotoxic and regulatory T cell molecule, cytotoxic and regulatory T-cell molecule
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CRTAM
Human CRTAM
0.1 mL
Primary
56253
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit