Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTAM Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550214

Catalog No. PA550214


Add to cart

Description

Description

The antibody detects endogenous levels of total CRTAM protein.

The CRTAM gene is upregulated in CD4-positive and CD8-positive T cells and encodes a type I transmembrane protein with V and C1-like Ig domains.
Specifications

Specifications

CRTAM
Polyclonal
Unconjugated
CRTAM
Liquid
CRTAM
Fusion protein corresponding to a region derived from internal residues of human cytotoxic and regulatory T cell molecule
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.6 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
O95727
CD355
Rabbit
IgG
100 μL
-20°C
Primary
56253
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit