Learn More
Abnova™ CRY1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001407T01
Catalog No. 89-940-608
Add to cart
Specifications
Specifications
Western Blot
293T
NM_004075.2
pCMV-CRY1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
CRY1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 66.4kDa
Transient overexpression cell lysate was tested with anti-ETS2antibody (Mfr. No. H00001407-B01) byWesternBlots.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)