Learn More
Aviva Systems Biology Corp CRY2 Antibody - N-terminal region (ARP52398_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52398_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRY2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CRY2. Alternate names: FLJ10332, HCRY2, KIAA0658, PHLL2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-63016