Learn More
Aviva Systems Biology Corp CRYBA1 Antibody - N-terminal region (ARP52061_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52061_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRYBA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CRYBA1. Alternate names: CRYB1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60678