Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYBB3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549023

Catalog No. PA549023

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Crystallins are the domit structural components of the vertebrate eye lens.
Specifications

Specifications

CRYBB3
Polyclonal
PBS with 0.09% sodium azide
P26998
Beta-crystallin B3, Beta-B3 crystallin, Beta-crystallin B3, N-terminally processed, CRYBB3, CRYB3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1417
Western Blot
Unconjugated
CRYBB3
Liquid
CRYBB3
KLH conjugated synthetic peptide between 179-207 amino acids from the C-terminal region of human CRYBB3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit