Learn More
Abnova™ CRYGS 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00001427T01
Catalog No. 89-940-650
Add to cart
Specifications
Specifications
Western Blot
293T
NM_017541
pCMV-CRYGS full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
CRYGS 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 21kDa
Transient overexpression cell lysate was tested with anti-CRYGS antibody (Mfr. No. H00001427-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)