Learn More
Tempshield™ Cryo-Gloves™ Blue Shoulder-Length Cryogenic Gloves
State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
$559.00 - $5460.00
Specifications
Certifications/Compliance (Europe) | EN 420, EN 511 |
---|---|
Description | Shoulder Length |
Color | Blue |
Hand | Specific |
Product Type | Cryogenic Gloves |
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
11-394-308
|
Tempshield™
SHL |
Large |
Pair for $559.00
Case of 10 PR for $5,320.00
|
|
|||||
19-159178
|
Tempshield™
SHXL |
X-Large |
Case of 10 for $5,460.00
|
|
|||||
Description
State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Blue | |
Cryogenic Gloves | |
Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems | |
-320°F | |
24.5 to 27.3 in. |
Shoulder Length | |
Specific | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
62 to 69.5 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.