Learn More
Thermo Scientific™ Cryo Gloves Available on GSA/VA Contract for Federal Government customers only.
Handle samples from ultra-low and cryogenic storage with these cryo gloves which offer excellent thermal protection.
Manufacturer:  Thermo Scientific™ 189565
 View more versions of this product
Catalog No. 13-994-015
  Check Availability
Add to cart
Specifications
Description
Handle samples from ultra-low and cryogenic storage with Thermo Scientific™ Cryo Gloves, which offer excellent thermal protection.
  • Comfortable, lightweight, breathable protection
  • With side release buckles for quick on/off
  • Available in a variety of sizes
Description & Specifications
Specifications
Large
Large Cryo Gloves
18 in.
45.72cm