Thermo Scientific™ Cryo Gloves Available on GSA/VA Contract for Federal Government customers only.
Handle samples from ultra-low and cryogenic storage with these cryo gloves which offer excellent thermal protection.
Manufacturer: Thermo Scientific™ 189563
 View more versions of this product
Catalog No. 13-994-013
  Check Availability
Add to Cart
    Specifications
    Type X-Large Cryo Gloves
    Length (English) 14 in.
    Length (Metric) 35.56cm
    Size X-Large
    Handle samples from ultra-low and cryogenic storage with Thermo Scientific™ Cryo Gloves, which offer excellent thermal protection.
    • Comfortable, lightweight, breathable protection
    • With side release buckles for quick on/off
    • Available in a variety of sizes
  • Description & Specifications
Related Products
Product Content Feedback