Learn More
Research Products International CorpSupplier Diversity Partner Cryostore Vial with O-Ring Seal, Flat Top Screw Cap Pre-Attached, Sterile, 2.0ml Capacity, 500 per Case DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 212406
Perfect for cryogenic work, these vials come with seals in a flat top screw cap which provides a writing surface. Caps are loosely pre-attached to vials to ensure product sterility. Freestanding polypropylene vials feature white printed graduations and marking area.
Catalog No. 50-476-502