Learn More
Thermo Scientific™ CryoPlus™ Temperature Recorder
Record seven days of temperature performance using the Thermo Scientific™ CryoPlus™ Temperature Recorder, for use with CryoPlus™ LN2 cryopreservation storage systems.
$2260.00
Specifications
For Use With (Equipment) CryoPlus 1,2,3,4
View More Specs
Products 1
Catalog Number Mfr. No. Voltage Price Quantity & Availability  
Catalog Number Mfr. No. Voltage Price Quantity & Availability  
13-262-795
View Documents
Thermo Scientific™
201280
220 V Each for $2,260.00
Add to cart
Description
Description
Record seven days of temperature performance using the Thermo Scientific™ CryoPlus™ Temperature Recorder, for use with CryoPlus LN2 cryopreservation storage systems.
Features:
  • 6 in. (15.2cm) dia.
  • With single pen and chart drive
  • -100°C to -200°C temperature range
Ordering Information:
Custom installed; contact your Sales Representative to specify top, left side or right side mount.
  • Warranty varies by country, please contact us for details
  • Specifications
    Specifications
    CryoPlus 1,2,3,4