Thermo Scientific™ Cryotome™ Cryostat Accessories
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Thermo Scientific™ Cryotome™ Cryostat Accessories.
Manufacturer: Thermo Scientific™ A78910057
 View more versions of this product
Catalog No. A78910057
  Check Availability
Add to Cart
    Specifications
    Description Cryocassettes, 28 x 40mm crosshatched (4/pk.)
    Item Description Cryocassettes, 28 x 40mm crosshatched
    Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Thermo Scientific™ Cryotome™ Cryostat Accessories.
    • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
    • Freezing time is shortened if freezing stage and tissue are placed on active Peltier
    • Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples
  • Description & Specifications
Related Products
Product Content Feedback