Learn More
Thermo Scientific™ Cryotome™ Cryostat Accessories
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Thermo Scientific™ Cryotome™ Cryostat Accessories.
Manufacturer:  Thermo Scientific™ A78930200
 View more versions of this product
Catalog No. A78930200
  Check Availability
Add to cart
Specifications
Description
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Thermo Scientific™ Cryotome™ Cryostat Accessories.
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on active Peltier
  • Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples
Description & Specifications
Specifications
Glass Anti-roll Plate
Glass Anti-Roll Plate
Glass anti-roll plate
FSE/FE/SME/E
FSE/FE/SME/E