Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ Cryptotanshinone

STAT3 inhibitor. Also displays multiple other activities

Manufacturer:  Tocris™ 3713/50

 View more versions of this product

Catalog No. 37-135-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Major tanshinone isolated from Salvia miltiorrhiza that exhibits multiple activities. Exhibits antitumor activity via inhibition STAT3 activity (IC50 = 4.6µM). Displays antibacterial and anti-inflammatory activity and acts as an antidiabetes and antiobesity agent via activation of AMP-activated protein kinase (AMPK). Also improves cognitive impairment in Alzheimer's disease transgenic mice by inhibition of acetylcholinesterase (IC50 = 4.09µM) and reduction in Aβ peptide generation.
  • Chemical Name: 1,2,6,7,8,9-Hexahydro-1,6,6-trimethyl[1,2-b]furan-10,11-dione
  • Formula: C19H20O3
  • CAS Number: 35825-57-1
  • Solubility: Soluble to 10mM in DMSO
  • Purity: >98
Description & Specifications

Specifications

Cryptotanshinone
>98
50mg
Cryptotanshinone
296.36
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit