Learn More
Crystal Models
Durable, permanently bonded crystalline models.
$227.00
Specifications
Type | Crystal molecular model |
---|---|
For Use With (Application) | 3-D lattice structure of crystal |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
S194365
![]() |
Denoyer Geppert Company
FOM512 |
Quartz |
Each for $227.00
|
|
S194364
![]() |
Denoyer Geppert Company
FOM530 |
Salt |
Each for $227.00
|
|
S194366
![]() |
Denoyer Geppert Company
FOM520 |
Ice |
Each for $227.00
|
|
Description
- Show the three-dimensional lattice structure of each crystal
- Visualize and explain properties of substances, concept of unit cells, and interatomic symmetry
- Different colored spheres are used to represent crystal structures containing more than one type of atom
- Diamond model is the most complex with 84 spheres
Specifications
Crystal molecular model | |
3-D lattice structure of crystal |