Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Crystal violet, ACS reagent

Catalog No. AC447572500
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle

CAS: 548-62-9 | C25H30ClN3 | 407.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Crystal violet
CAS 548-62-9, 90-94-8
Melting Point 173°C
Packaging Glass bottle
Molecular Formula C25H30ClN3
Quantity 250 g
Merck Index 15,443
Synonym crystal violet, gentian violet, basic violet 3, methylrosaniline chloride, aniline violet, hexamethyl violet, gentiaverm, hexamethylpararosaniline chloride, pyoktanin, adergon
Solubility Information Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform,1g/10 mL alcohol,1g/15 mL glycerin,practically insoluble in ether
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
Molecular Weight (g/mol) 407.99
PubChem CID 11057
ChEBI CHEBI:41688
Formula Weight 407.99
Grade ACS Reagent
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.