Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet TS 1% in Glacial Acetic Acid, Reagents

Catalog No. p-200021563
missing translation for 'orderingAttributeHoverText'
Quantity:
120 mL
500 mL

Crystal Violet TS 1% in Glacial Acetic Acid, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-19-7, 548-62-9
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Gentian Violet, Basic Violet 3
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Boiling Point 118°C
Percent Purity 1%
Grade USP
Packaging Glass Bottle
Specific Gravity 1.05
Color Purple
pH <2
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Crystal Violet TS 1% in Glacial Acetic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.