Learn More
Hampton Research CRYSTALEX 96 WELL PLATE 10CSC
SDP
Supplier:  Hampton Research HR8160
Encompass
HR8-160/CrystalEX Second Generation 96 Well Plate (Corning 3557), 1 ul conical flat drop well, 5 drop well, Pzero - 10 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-020-0