Learn More
Crystalgen Bio-Degradable Green Culture Flasks
$212.00 - $296.00
Specifications
Material Plastic
View More Specs
Products 8
Catalog Number Mfr. No. Bottom Shape Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Bottom Shape Capacity (Metric) Price Quantity & Availability  
05-539-805 View Documents Crystalgen
260002
Standard 250 mL Case of 24 for $296.00
Add to cart
05-539-804 View Documents Crystalgen
260001
Standard 125 mL Case of 24 for $235.00
Add to cart
05-539-806 View Documents Crystalgen
260003
Standard 500 mL Case of 12 for $212.00
Add to cart
05-539-808 View Documents Crystalgen
260005
Baffled 125 mL Case of 24 for $235.00
Add to cart
05-539-807 View Documents Crystalgen
260004
Standard 1000 mL Case of 12 for $271.50
Add to cart
05-539-809 View Documents Crystalgen
260006
Baffled 250 mL Case of 24 for $296.00
Add to cart
05-539-810 View Documents Crystalgen
260007
Baffled 500 mL Case of 12 for $212.00
Add to cart
05-539-811 View Documents Crystalgen
260008
Baffled 1000 mL Case of 12 for $296.00
Add to cart
Description
Description
100% bio-degradable lab disposable plastic culture flasks
  • Made of raw polymer that can be degraded by the soil within a year after disposal or 70 days under composting conditions
  • Certified by LNE EN13432 biodegradable report and FDA Biomedical Applications
Available in Standard or Baffled
All shaking cultures, mixing, and storage of biomedical fluids
Specifications
Specifications
Plastic