Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crystallized Bovine Albumin, Thermo Scientific Chemicals
Crystallized Bovine Albumin
Supplier: Thermo Scientific J1085622
Description
- Source: Bovine blood
- Assay (electrophoresis): ≥99%
- Identity: By IR
- Protein: ≥98%
- pH (7%, 0.15M NaCl): 5.0 - 5.4
- Moisture (KF) when packaged: %le;5.0%
- Sodium (Na): 0 - 5.0mg/g
- Chloride (Cl): 0 - 3.0mg/g
- CAS: 9048-46-8
Specifications
5.0 to 5.4 | |
Bovine blood | |
Bovine Albumin |
100 g | |
Plastic bottle | |
Crystalline |
Safety and Handling
EINECSNumber : 232-936-2
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction