Learn More
Hampton Research CRYSTALQUICK 96 WELL PL 10CSC
SDP
Supplier:  Hampton Research HR3283
Encompass
HR3-283/CrystalQuick 96 Well Sitting Drop Plate (Greiner 609120), 2 ul round drop well, 3 drop well - 10 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-018-9