Learn More
Hampton Research CRYSTALQUICK PLUS 96 WE 80CSC
SDP
Supplier:  Hampton Research HR3093G
Encompass
HR3-093G/CrystalQuick Plus 96 Well Sitting Drop Plate (Greiner 609180), 4 ul square drop well, 1 drop well, LBR, hydrophobic, low profile - 80 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-053-2
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.