Learn More
CRYZL1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 168941AP
Catalog No. 50-173-1242
Add to cart
Description
Description
This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.
Specifications
Specifications
CRYZL1
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95825, Q921W4
4P11, CRYZL1, Protein 4P11, QOH 1, Zeta crystallin homolog
Rabbit
IgG
150 μL
-20°C
Primary
288256, 66609, 9946
Western Blot
Unconjugated
CRYZL1
Liquid
Cryzl1
CRYZL1 Fusion Protein Ag10419
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat