Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYZL1 Mouse anti-Human, Clone: OTI2C3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525971

Catalog No. PIMA525971


Add to cart

Description

Description

This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.
Specifications

Specifications

CRYZL1
Monoclonal
0.77 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O95825
crystallin, zeta (quinone reductase)-like 1, 4P11, QOH-1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9946
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2C3
Unconjugated
CRYZL1
Liquid
CRYZL1
Full length human recombit protein of CRYZL1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit