Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CS 2100, Tocris Bioscience™
GREENER_CHOICE

Catalog No. p-4816717
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective S1P1 agonist

Sphingosine-1-phosphate receptor 1 (S1P1) agonist (EC50 = 4.0 nM). Exhibits 5000-fold selectivity for human S1P1 over S1P3. Displays efficacy in a rat adjuvant-induced arthritis model.

Chemical Identifiers

CAS 913827-99-3
Molecular Formula C25H23N3O4S
Molecular Weight (g/mol) 461.536
InChI Key DWVJASHDNJMDNH-UHFFFAOYSA-N
Synonym 1-4-ethyl-5-5-4-phenoxyphenyl-1,2,4-oxadiazol-3-yl-2-thienyl methyl azetidine-3-carboxylic acid, 1-4-ethyl-5-5-4-phenoxyphenyl-1,2,4-oxadiazol-3-yl-2-thienyl methyl-3-azetidinecarboxylic acid, 1-4-ethyl-5-5-4-phenoxyphenyl-1,2,4-oxadiazole-3-yl-2-thienyl methyl-3-azetidinecarboxylic acid
PubChem CID 11977938
IUPAC Name 1-[[4-ethyl-5-[5-(4-phenoxyphenyl)-1,2,4-oxadiazol-3-yl]thiophen-2-yl]methyl]azetidine-3-carboxylic acid
SMILES CCC1=C(SC(=C1)CN2CC(C2)C(=O)O)C3=NOC(=N3)C4=CC=C(C=C4)OC5=CC=CC=C5

Specifications

Quantity 10 mg
Formula Weight 461.53
Percent Purity >97%
Chemical Name or Material CS 2100
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.