Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Cs/Ba-137 Isotope Generator Products for Science Education
Ideal for radioactive decay experiments.
$22.81 - $373.68
Specifications
For Use With (Application) Radioactive decay experiment
View More Specs
Includes: Generator, syringe, tube, 250mL of solution, and a convenient storage case.
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S83190 Products for Science Education
View Documents
Spectrum Techniques IncSupplier Diversity Partner
ISO
Isotope generator kit
Each for $373.68
N/A
S83191 Products for Science Education
View Documents
Spectrum Techniques IncSupplier Diversity Partner
ELSN
Extra eluting solution
Each for $22.81
N/A
Description
Description
This CS-137/Ba-137m Isotope Generator was designed for school experiments to demonstrate the properties of radioactive decay. Each generator contains 10uCi of Cs-137.
Parent CS-137 isotope
  • Half-life of 30.1 years
  • Beta decays to the metastable state of Ba-137m
  • Further decays by gamma emission with a half-life of 2.6 minutes to the stable Ba-137 element
  • Ba-137m is milked from the generator, leaving behind the parent Cs-137
Ba-137 solution
  • Used to perform exponential decay experiments
  • After a 30-minute class, the short half-life of the solution reduces the residual activity, to less than 1/1000th of its initial activity
  • Safe for disposal by pouring down the drain
Order Info
This generator consists of only one exempt quantity, making it free from specific state and NRC licensing.
Specifications
Specifications
Radioactive decay experiment