Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CS Citrate Synthase Mouse anti-Human, Mouse, Rat, Zebrafish, DyLight 650, Clone: 1761, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243648C

Catalog No. NBP243648C


Add to cart

Description

Description

CS Citrate Synthase Monoclonal antibody specifically detects CS Citrate Synthase in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CS Citrate Synthase
1761
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
CS
Recombinant protein encompassing a sequence within the center region of human Citrate synthetase. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat, Zebrafish
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
citrate synthase, citrate synthase, mitochondrial, EC 2.3.3, EC 2.3.3.1
Mouse
IgG2a
0.1 ml
Primary
1431
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit