Learn More
CSAFE LLC CSafe Global 5L, 96hr, 2°C to 8°C (Refrigerated) Parcel Shipper - ISTA 7D Qualified - Vacuum Insulation Panel (VIP) Technology. Perfect for long duration shipments of Vaccines, Biologics and refrigerated therapies; including COVID-19 shipments
Supplier:  CSAFE LLC A05R096000U
Encompass_Preferred
This item has a minimum order quantity of 36 per supplier requirements
Temperature-controlled parcel shipper solutions are constructed using best-in-class Vacuum Insulated Panel (VIP) technology, assuring greater peace of mind in your cold chain. These shippers provide industry-leading temperature management performance for the safe transport of vital life-science products such as Vaccines, Biologics, Cell Therapies, and Oncology Treatments to distribution centers, healthcare facilities and direct to patients.This shipping solution has a 5L payload, qualified to maintain a strict 2°C to 8°C temp range (refrigerated) for 96 hours against ISTA 7D’s worst-case challenge testing. CSafe utilizes proprietary, single-temperature Phase Change Material (PCM) for a simplistic, universal pack-out. This system has also passed ISTA 3A Distribution Testing.This 35L solution is further protected by its double-wall outer corrugate, making this shipper ideal as a reusable system
Catalog No. NC1866792
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.