Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

csBAP31 Mouse anti-Human, Unconjugated, Clone: SP1-B7, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen csBAP31
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC17121MI
View Documents
MilliporeSigma
MABC1712100UL
100 μL
Each for $386.00
Add to cart
 
MABC17122MI
View Documents
MilliporeSigma
MABC171225UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

csBAP31 Monoclonal antibody specifically detects csBAP31 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

csBAP31
Unconjugated
Mouse
Cell surface B-cell receptor-associated protein 31;BCR-associated protein 31;Bap31;6C6-AG tumor-associated antigen;Protein CDM;p28
A549 human carcinoma sphere cells.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Flow Cytometry, Immunoprecipitation, Western Blot
Purified
Human
BCAP31, BAP31, DXS1357E
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit