Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSE1L/CAS/Exportin-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238384
Description
CSE1L/CAS/Exportin-2 Polyclonal specifically detects CSE1L/CAS/Exportin-2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CSE1L/CAS/Exportin-2 | |
Polyclonal | |
Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
P55060 | |
CSE1L | |
This antibody was developed against a recombinant protein corresponding to amino acids: HGITQANELVNLTEFFVNHILPDLKSANVNEFPVLKADGIKYIMIFRNQVPKEHLLVSIPLLINHLQAESIVVHTYAAHALERLFTMRGPNNATLF | |
0.1 mL | |
Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers | |
1434 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CASMGC117283, Cellular apoptosis susceptibility protein, chromosome segregation 1 (yeast homolog)-like, Chromosome segregation 1-like protein, CSE1, CSE1 chromosome segregation 1-like (yeast), CSE1L, Exp2, exportin-2, Importin-alpha re-exporter, MGC130037, XPO2MGC130036 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction