Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSE1L/CAS/Exportin-2 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: 729, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243610IR

Catalog No. NBP243610IR


Add to cart

Description

Description

CSE1L/CAS/Exportin-2 Monoclonal antibody specifically detects CSE1L/CAS/Exportin-2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

CSE1L/CAS/Exportin-2
729
Western Blot
Purified
CSE1L
Recombinant protein encompassing a sequence within the center region of human CSE1L. The exact sequence is proprietary.
Protein G purified
Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers
Primary
1434
Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CASMGC117283, Cellular apoptosis susceptibility protein, chromosome segregation 1 (yeast homolog)-like, Chromosome segregation 1-like protein, CSE1, CSE1 chromosome segregation 1-like (yeast), CSE1L, Exp2, exportin-2, Importin-alpha re-exporter, MGC130037, XPO2MGC130036
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit