Learn More
CSF1 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29930
Catalog No. 89-033-218
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human CSF1.
  • High specificity
  • High reactivity
Specifications
Specifications
CSF1
Unconjugated
Immunofluorescence (10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot The optimal working dilution should be determined by the end user.
NM_000757
MCSF|MGC31930
Rabbit
Affinity purification
RUO
Primary
1435
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human CSF1.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
CSF1
A synthetic peptide corresponding to internal region of human CSF1.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)