Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ CSF1R (FMS) Recombinant Human Protein
$540.00
Specifications
Accession Number | NP_005202 |
---|---|
Molecular Weight | 44.8 kDa |
Gene Alias | CSF1R (FMS), FMS, CSFR, c-FMS, CD115 |
KinaseFamily | PDGFR Kinase Family |
Recombinant | Yes |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
PV3249
![]() |
Thermo Scientific™
PV3249 |
10μg | 10μg |
Each of 1 for $540.00
|
|
Description
CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias.Specifications
NP_005202 | |
CSF1R (FMS), FMS, CSFR, c-FMS, CD115 | |
Yes | |
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinase Assay |
44.8 kDa | |
PDGFR Kinase Family | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title