Learn More
CSF2RA 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00001438T01
Catalog No. 89-008-831
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
CSF2RA 293T Cell Transient Overexpression Lysate(Denatured)
46.2
pCMV-CSF2RA full-length
100 μL
Human
293T
NM_006140
6.8
Transient overexpression cell Lysate was tested with Anti-CSF2RA antibody (H00001438-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.