Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSHL1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298959MFV610
Description
CSHL1 Polyclonal antibody specifically detects CSHL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CSHL1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
chorionic somatomammotropin hormone-like 1, Chorionic somatomammotropin-like, CS-5, CSHP1, CSLchorionic somatomammotropin CS-5, growth hormone cluster, hCS-L, Lactogen-like, MGC149868 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CSHL1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
1444 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction