Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSHL1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548503

Catalog No. PA548503


Only null left
Add to Cart

Description

Description

CSHL1 Polyclonal Antibody for Western Blot, IHC (P)

May be a novel gestational hormone required to compensate for absence of other members of the GH/CS cluster during gestation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CSHL1
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q14406
CSHL1
KLH conjugated synthetic peptide between 156-185 amino acids from the C-terminal region of human CSHL1.
400 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CSHL1
chorionic somatomammotropin CS-5; chorionic somatomammotropin hormone like 1; chorionic somatomammotropin hormone-like 1; Chorionic somatomammotropin-like; CS-5; CSHL1; CSHP1; CSL; GHB4; growth hormone B4; growth hormone cluster; hCS-L; Lactogen-like
Rabbit
Antigen affinity chromatography, Protein A
RUO
1444
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.